EMAIL THIS PAGE TO A FRIEND
Biotechnology journal
Editorial: Biotechnology Journal 10 year Anniversary - Thank you for your continued support.
PMID 26663833
Abstract
When we came to realize that Biotechnology Journal is 10 years "old" and she is no longer "young", we started to look back and look forward again. We would like to thank all of the editors, authors, reviewers, readers and partners for your continued support during the past ten years!