EMAIL THIS PAGE TO A FRIEND
Mediators of inflammation
Altered Innate Immune Responses in Neutrophils from Patients with Well- and Suboptimally Controlled Asthma.
PMID 26663987
Abstract
Respiratory infections are a major cause of asthma exacerbations where neutrophilic inflammation dominates and is associated with steroid refractory asthma. Structural airway cells in asthma differ from nonasthmatics; however it is unknown if neutrophils differ. We investigated neutrophil immune responses in patients who have good (AGood) and suboptimal (ASubopt) asthma symptom control. Peripheral blood neutrophils from AGood (ACQ < 0.75, n = 11), ASubopt (ACQ > 0.75, n = 7), and healthy controls (HC) (n = 9) were stimulated with bacterial (LPS (1 μg/mL), fMLF (100 nM)), and viral (imiquimod (3 μg/mL), R848 (1.5 μg/mL), and poly I:C (10 μg/mL)) surrogates or live rhinovirus (RV) 16 (MOI1). Cell-free supernatant was collected after 1 h for neutrophil elastase (NE) and matrix metalloproteinase- (MMP-) 9 measurements or after 24 h for CXCL8 release. Results. Constitutive NE was enhanced in AGood neutrophils compared to HC. fMLF stimulated neutrophils from ASubopt but not AGood produced 50% of HC levels. fMLF induced MMP-9 was impaired in ASubopt and AGood compared to HC. fMLF stimulated CXCL8 but not MMP-9 was positively correlated with FEV1 and FEV1/FVC. ASubopt and AGood responded similarly to other stimuli. Circulating neutrophils are different in asthma; however, this is likely to be related to airflow limitation rather than asthma control.
Related Materials