EMAIL THIS PAGE TO A FRIEND
Food microbiology
Inactivation kinetics of Listeria monocytogenes and Salmonella enterica serovar Typhimurium on fresh-cut bell pepper treated with slightly acidic electrolyzed water combined with ultrasound and mild heat.
PMID 26678144
Abstract
The goal of this study was to enhance the antimicrobial effect of slightly acidic electrolyzed water (SAEW) through addition of synergistic treatment with ultrasound (US) and mild heat treatment in order to improve the microbial safety of fresh-cut bell pepper. To evaluate the synergistic effects, the Weibull model was used to mathematically measure the effectiveness of the individual and combined treatments against Listeria monocytogenes and Salmonella Typhimurium on the pepper. The combined treatment (SAEW+US+60xa0°C) resulted in the TR values of 0.04 and 0.09xa0min for L.xa0monocytogenes and S.xa0Typhimurium, respectively, as consequence of the minimum value. Subsequently, texture analysis was carried out to test the potential effect on quality of the samples due to the involved mild heat and ultrasound treatment. When compared to the control, there was no significant change (pxa0≥xa00.05) in the texture (color and hardness) of the samples that were treated by 1xa0min of the combined treatment (SAEW+US+60xa0°C) during storage at 4xa0°C for 7 days. This combined treatment achieved approximately 3.0 log CFU/g reduction in the two pathogens. The results demonstrate that the involved hurdle factors which are ultrasound and mild heat achieved the synergistic effect of SAEW against the two pathogens. According to the results of texture analysis, 1xa0min of SAEW+US+60xa0°C is the optimal condition due to without negative influence on the quality of the samples during the storage. The optimal condition shows the enhanced antimicrobial effect of SAEW and enables to improve microbial safety of fresh bell pepper in food industry as a consequence of hurdle approach.