EMAIL THIS PAGE TO A FRIEND
Genome announcements
Draft Genome Sequence of Thermophilic Geobacillus sp. Strain Sah69, Isolated from Saharan Soil, Southeast Algeria.
PMID 26679578
Abstract
Geobacillus spp. are potential sources of novel enzymes, such as those involved in the degradation of recalcitrant polymers. Here, we report a Geobacillus genome that may help reveal genomic differences between this strain and publicly available representatives of the same genus from diverse niches.