EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Cyclin A2 and CDK2 as Novel Targets of Aspirin and Salicylic Acid: A Potential Role in Cancer Prevention.
PMID 26685215
Abstract
Data emerging from the past 10 years have consolidated the rationale for investigating the use of aspirin as a chemopreventive agent; however, the mechanisms leading to its anticancer effects are still being elucidated. We hypothesized that aspirin's chemopreventive actions may involve cell-cycle regulation through modulation of the levels or activity of cyclin A2/cyclin-dependent kinase-2 (CDK2). In this study, HT-29 and other diverse panel of cancer cells were used to demonstrate that both aspirin and its primary metabolite, salicylic acid, decreased cyclin A2 (CCNA2) and CDK2 protein and mRNA levels. The downregulatory effect of either drugs on cyclin A2 levels was prevented by pretreatment with lactacystin, an inhibitor of proteasomes, suggesting the involvement of 26S proteasomes. In-vitro kinase assays showed that lysates from cells treated with salicylic acid had lower levels of CDK2 activity. Importantly, three independent experiments revealed that salicylic acid directly binds to CDK2. First, inclusion of salicylic acid in naïve cell lysates, or in recombinant CDK2 preparations, increased the ability of the anti-CDK2 antibody to immunoprecipitate CDK2, suggesting that salicylic acid may directly bind and alter its conformation. Second, in 8-anilino-1-naphthalene-sulfonate (ANS)-CDK2 fluorescence assays, preincubation of CDK2 with salicylic acid dose-dependently quenched the fluorescence due to ANS. Third, computational analysis using molecular docking studies identified Asp145 and Lys33 as the potential sites of salicylic acid interactions with CDK2. These results demonstrate that aspirin and salicylic acid downregulate cyclin A2/CDK2 proteins in multiple cancer cell lines, suggesting a novel target and mechanism of action in chemoprevention. Biochemical and structural studies indicate that the antiproliferative actions of aspirin are mediated through cyclin A2/CDK2.