EMAIL THIS PAGE TO A FRIEND
Neural regeneration research
A novel therapeutic target for peripheral nerve injury-related diseases: aminoacyl-tRNA synthetases.
PMID 26692865
Abstract
Aminoacyl-tRNA synthetases (AminoARSs) are essential enzymes that perform the first step of protein synthesis. Beyond their original roles, AminoARSs possess non-canonical functions, such as cell cycle regulation and signal transduction. Therefore, AminoARSs represent a powerful pharmaceutical target if their non-canonical functions can be controlled. Using AminoARSs-specific primers, we screened mRNA expression in the spinal cord dorsal horn of rats with peripheral nerve injury created by sciatic nerve axotomy. Of 20 AminoARSs, we found that phenylalanyl-tRNA synthetase beta chain (FARSB), isoleucyl-tRNA synthetase (IARS) and methionyl-tRNA synthetase (MARS) mRNA expression was increased in spinal dorsal horn neurons on the injured side, but not in glial cells. These findings suggest the possibility that FARSB, IARS and MARS, as a neurotransmitter, may transfer abnormal sensory signals after peripheral nerve damage and become a new target for drug treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049234
Anti-AIF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA036678
Anti-FARSB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701217
Anti-FARSB antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1409302
Anti-MARS antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004125
Anti-MARS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101437
Anti-MARS antibody produced in rabbit, affinity isolated antibody
SAB2107539
Anti-SLA2 antibody produced in rabbit, affinity isolated antibody
SAB2107540
Anti-SLA2 antibody produced in rabbit, affinity isolated antibody
SAB1411502
Anti-SLA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV46359
Anti-TARS antibody produced in rabbit, IgG fraction of antiserum
HPA037425
Anti-TARS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2702096
Anti-TARS antibody produced in rabbit
D2510
Deoxycholic acid, ≥98% (HPLC)
C24H40O4
D4297
Deoxycholic acid, BioXtra, ≥98% (HPLC)
C24H40O4
30960
Deoxycholic acid, ≥99.0% (T)
C24H40O4
WH0000199M1
Monoclonal Anti-AIF1 antibody produced in mouse, clone 2A2-B6, purified immunoglobulin, buffered aqueous solution
WH0010056M1
Monoclonal Anti-FARSLB antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution
WH0004141M1
Monoclonal Anti-MARS antibody produced in mouse, clone 5G5, purified immunoglobulin, buffered aqueous solution
WH0084174M1
Monoclonal Anti-SLA2 antibody produced in mouse, clone 3G5, purified immunoglobulin, buffered aqueous solution
SAB4100503
Monoclonal Anti-SLA2 antibody produced in mouse, clone SLA2-A443, culture supernatant
WH0006897M1
Monoclonal Anti-TARS antibody produced in mouse, clone 1A9, purified immunoglobulin, buffered aqueous solution
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
74032
Nitrotetrazolium Blue chloride, ≥98.0% (calc. based on dry substance, T)
C40H30Cl2N10O6
P7626
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
C7H7FO2S
78830
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
C7H7FO2S