EMAIL THIS PAGE TO A FRIEND
Neural regeneration research
The HMGB1 signaling pathway activates the inflammatory response in Schwann cells.
PMID 26692874
Abstract
Schwann cells are not only myelinating cells, but also function as immune cells and express numerous innate pattern recognition receptors, including the Toll-like receptors. Injury to peripheral nerves activates an inflammatory response in Schwann cells. However, it is unclear whether specific endogenous damage-associated molecular pattern molecules are involved in the inflammatory response following nerve injury. In the present study, we demonstrate that a key damage-associated molecular pattern molecule, high mobility group box 1 (HMGB1), is upregulated following rat sciatic nerve axotomy, and we show colocalization of the protein with Schw-ann cells. HMGB1 alone could not enhance expression of Toll-like receptors or the receptor for advanced glycation end products (RAGE), but was able to facilitate migration of Schwann cells. When Schwann cells were treated with HMGB1 together with lipopolysaccharide, the expression levels of Toll-like receptors and RAGE, as well as inflammatory cytokines were upregulated. Our novel findings demonstrate that the HMGB1 pathway activates the inflammatory response in Schwann cells following peripheral nerve injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3262
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
C10H12N5O6P · C4H11NO3
F8775
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CH2O
F8923
Formaldehyde solution, meets USP testing specifications, contains 9.0-15% methanol as stabilizer
CH2O
252549
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
CH2O
F15587
Formaldehyde solution, 37 wt. % in H2O
CH2O
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
33220
Formaldehyde solution, puriss. p.a., ACS reagent, ≥36.5%
CH2O
15512
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
CH2O
47083-U
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
CH2O
606758
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
12CH2O
H4652 HMG-1 human, lyophilized powder, ≥90% (SDS-PAGE), Histidine-tagged, recombinant, expressed in E. coli
74255
Sodium dodecyl sulfate, dust-free pellets, ≥99.0% (GC)
C12H25NaO4S
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S
71717
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
C12H25NaO4S
62862
Sodium dodecyl sulfate, ≥90%
C12H25NaO4S
75746
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
C12H25NaO4S
71736
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
C12H25NaO4S
05030
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
C12H25NaO4S