EMAIL THIS PAGE TO A FRIEND
Talanta
Development of an analytical method for the targeted screening and multi-residue quantification of environmental contaminants in urine by liquid chromatography coupled to high resolution mass spectrometry for evaluation of human exposures.
PMID 26695319
Abstract
The aim of this study was to develop an analytical method and contribute to the assessment of the Exposome. Thus, a targeted analysis of a wide range of contaminants in contact with humans on daily routines in urine was developed. The method focused on a list of 38 contaminants, including 12 pesticides, one metabolite of pesticide, seven veterinary drugs, five parabens, one UV filter, one plastic additive, two surfactants and nine substances found in different products present in the everyday human environment. These contaminants were analyzed by high performance liquid chromatography coupled to high resolution mass spectrometry (HPLC-HRMS) with a quadrupole-time-of-flight (QqToF) instrument from a raw urinary matrix. A validation according to the FDA guidelines was employed to evaluate the specificity, linear or quadratic curve fitting, inter- and intra-day precision, accuracy and limits of detection and quantification (LOQ). The developed analysis allows for the quantification of 23 contaminants in the urine samples, with the LOQs ranging between 4.3 ng.mL(-1) and 113.2 ng.mL(-1). This method was applied to 17 urine samples. Among the targeted contaminants, four compounds were detected in samples. One of the contaminants (tributyl phosphate) was detected below the LOQ. The three others (4-hydroxybenzoic acid, sodium dodecylbenzenesulfonate and O,O-diethyl thiophosphate potassium) were detected but did not fulfill the validation criteria for quantification. Among these four compounds, two of them were found in all samples: tributyl phosphate and the surfactant sodium dodecylbenzenesulfonate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W395900
2-Phenylphenol, ≥99%, FG
C12H10O
P28263
2-Phenylphenol, 99%
C12H10O
278564
4′-Hydroxyacetophenone, 99%
C8H8O2
442840
4,4′-Isopropylidenediphenol, analytical standard
C15H16O2
240141
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
C7H6O3
H20059
4-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
W398608
4-Hydroxybenzoic acid, ≥99%, FG
C7H6O3
237825
Bis(2-ethylhexyl) phosphate, 97%
C16H35O4P
68572
Dibutyl phosphate, ≥97.0% (T)
C8H19O4P
D2425
Diuron, ≥98%
C9H10Cl2N2O
111988
Ethyl 4-hydroxybenzoate, ReagentPlus®, 99%
C9H10O3
77249
Heptafluorobutyric acid, ≥99.0% (GC)
C4HF7O2
164194
Heptafluorobutyric acid, 98%
C4HF7O2
N12393
Methamidophos, analytical standard
C2H8NO2PS
PS676
Methamidophos, analytical standard
C2H8NO2PS
H5501
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
C8H8O3
H6654
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
C8H8O3
H3647
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
C8H8O3
M8911
Methyl 4-hydroxybenzoate, analytical standard
C8H8O3
W271004
Methyl 4-hydroxybenzoate, ≥99%, FCC
C8H8O3
47889
Methyl Paraben, analytical standard
C8H8O3
394459
Perfluorononanoic acid, 97%
C9HF17O2
171468
Perfluorooctanoic acid, 96%
C8HF15O2
396575
Perfluoropentanoic acid, 97%
C5HF9O2
158615
Tributyl phosphate, 97%
C12H27O4P
240494
Tributyl phosphate, ≥99%
C12H27O4P
29226
Undecafluorohexanoic acid, ≥97.0% (T)
C6HF11O2