EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Highly Efficient Planar Perovskite Solar Cells Via Interfacial Modification with Fullerene Derivatives.
PMID 26701816
Abstract
Planar heterojunction perovskite solar cells with a high efficiency up to 17.76% are fabricated by modifying the compact TiO2 (c-TiO2) with a [6,6]-phenyl-C61-butyric acid (PCBA) monolayer. High quality CH3NH3PbI3 films can be easily fabricated on PCBA-modified c-TiO2 substrates by a one-step solution processing method. Significant improvements of the device parameters are observed after PCBA modification. A high open-circuit voltage (Voc) of 1.16 V has been achieved, indicating that the PCBA monolayer can act as a hole blocking layer to reduce the trap site density atop the c-TiO2 and the hole recombination at the c-TiO2 /perovskite interface. The enhancement of the fill factor, as well as the partial quenching of the fluorescence of perovskite after modification with PCBA, reveals that the charge extraction is improved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
426466
Methylamine solution, 40 wt. % in H2O
CH5N