EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development of a monoclonal antibody-based indirect competitive enzyme-linked immunosorbent assay for nitroimidazoles in edible animal tissues and feeds.
PMID 26717017
Abstract
The misuse of nitroimidazoles (NDZs) can lead to NDZs residues in edible animal tissues, which would be harmful to consumer health. To quickly monitor NDZs residues in edible animal tissues and feed, a monoclonal antibody-based indirect competitive enzyme-linked immunosorbent assay (ic-ELISA) with a simple sample preparation method and clean-up was developed in the present study. At first, a broad-specificity monoclonal antibody, 1D5, against NDZs has been produced, which the IC50 values of the NDZs, dimetridazole, ipronidazole, ronidazole hydroxydimetridazole, and hydroxyipronidazole, were 4.79μgL(-1), 0.47μgL(-1), 5.97μgL(-1), 23.48μgL(-1), and 15.03μgL(-1), respectively. The limit of detection of the method for the NDZ matrix calibration ranged from 4.2μgkg(-1) to 50.3μgkg(-1) in the feed matrices and from 0.11μgkg(-1) to 4.11μgkg(-1) in the edible animal tissues matrices. The recoveries of the NDZs were in the range of 75.5-111.8%. The CVs were less than 14.4%. A good correlation (r=0.9905) between the ELISA and HPLC-MS results of the tissues demonstrated the reliability of the developed ic-ELISA, which makes it a useful tool for screening of NDZs in animal edible tissue and feed.