EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis, colon-targeted studies and pharmacological evaluation of an anti-ulcerative colitis drug 4-Aminosalicylic acid-β-O-glucoside.
PMID 26717200
Abstract
A glycoside prodrug of 4-aminosalicylic acid (4-ASA) with d-glucose was synthesized for targeted drug delivery to inflammatory bowel. The inxa0vitro assessment of 4-aminosalicylic acid-β-O-glucoside (4-ASA-Glu) as a colon-specific prodrug was studied using colitis rat with the healthy one as control. The stability studies in aqueous buffers (pH 1.2, 6.8 and 7.4) indicated that 4-ASA-Glu was stable over a period of 12xa0h. The incubation of 4-ASA-Glu with cecal or colonic contents of healthy rats at 37xa0°C released 4-ASA in 77 or 80% of the dose in 12xa0h, respectively. The amount of 4-ASA liberated from the incubation of 4-ASA-Glu in cecal or colonic contents of colitis rats at 37xa0°C was 69 or 79% in 12xa0h respectively, while less than 9% 4-ASA was detected from the incubation of 4-ASA-Glu with the homogenates of stomach or small intestine. The curative effect of 4-ASA-Glu was evaluated in 2, 4, 6-trinitrobenzenesulfonic acid (TNBS) induced experimental colitis model in male Sprague-Dawley (SD) rats. It was found that 4-ASA-Glu possess significantly ameliorate effect than sulfasalazine, oral 4- and 5-aminosalicylic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119938
Benzyl chloroformate, ≥98%
C8H7ClO2