EMAIL THIS PAGE TO A FRIEND
Scientific reports
Galangin sensitizes TRAIL-induced apoptosis through down-regulation of anti-apoptotic proteins in renal carcinoma Caki cells.
PMID 26725939
Abstract
Galangin, bioflavonoids, has been shown anti-cancer properties in various cancer cells. In this study, we investigated whether galangin could enhance TRAIL-mediated apoptosis in TRAIL resistant renal carcinoma Caki cells. Galangin alone and TRAIL alone had no effect on apoptosis, while combined treatment with galangin and TRAIL significantly induced apoptosis in renal carcinoma (Caki, ACHN and A498) but not normal cells (normal mouse kidney cells and human normal mesangial cells). Galangin induced down-regulation of Bcl-2 protein at the transcriptional level via inhibition of NF-κB activation but not p53 pathway. Furthermore, galangin induced down-regulation of cFLIP, Mcl-1 and survivin expression at the post-translational levels, and the over-expression of Bcl-2, cFLIP, Mcl-1 and survivin markedly reduced galangin-induced TRAIL sensitization. In addition, galangin increased proteasome activity, but galangin had no effect on expression of proteasome subunits (PSMA5 and PSMD4). In conclusion, our investigation suggests that galangin is a potent candidate for sensitizer of TRAIL resistant cancer cell therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282200
Galangin, autophagy inducing flavonoid
C15H10O5