EMAIL THIS PAGE TO A FRIEND
AMB Express
Effect of site-directed mutagenesis at the GGEEF domain of the biofilm forming GGEEF protein from Vibrio cholerae.
PMID 26728467
Abstract
Vibrio cholerae, the cause of seven noted pandemics, leads a dual lifecycle-one in the human host in its virulent form, and the other as a sessile, non-virulent bacterium in aquatic bodies in surface biofilms. Surface biofilms have been attributed to be associated with a ubiquitous protein domain present in all branches of bacteria, known as the GGD(/E)EF domain. While the diguanlyate cyclase activities of these proteins are universally established, the role of these proteins as diguanlyate-specific phosphodiesterases in conjunction with a EAL domain has also been reported. The VC0395_0300 protein from V. cholerae which shows biofilm forming abilities also acts as a phosphodiesterase. Interestingly, this GGD(/E)EF protein contains a EAL site in the reverse orientation. We attempted to mutate the GGEEF signature along the sequence by site-directed mutagenesis. The resultant mutants (Sebox5-7) did not show much difference in phosphodiesterase activity in comparison with the wild type protein (Sebox3), indicating the independence of the phosphodiesterase activity of the protein from the GGD(/E)EF domain. However, the ability of the mutants to form surface biofilm was significantly lesser in the case of mutations in the three central positions of the signature domain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123943
Bis(4-nitrophenyl) phosphate, 99%
C12H9N2O8P