PloS one
Calcium Signaling Is Required for Erythroid Enucleation.
PMID 26731108
Abstract
Although erythroid enucleation, the property of erythroblasts to expel their nucleus, has been known for 7ore than a century, surprisingly little is known regarding the molecular mechanisms governing this unique developmental process. Here we show that similar to cytokinesis, nuclear extrusion requires intracellular calcium signaling and signal transduction through the calmodulin (CaM) pathway. However, in contrast to cytokinesis we found that orthochromatic erythroblasts require uptake of extracellular calcium to enucleate. Together these functional studies highlight a critical role for calcium signaling in the regulation of erythroid enucleation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1619
CGS-9343B, ≥98% (HPLC), powder
C26H28N4O2 · C4H4O4
K4144
KB-R7943, ≥98% (HPLC), powder
C16H17N3O3S·CH3SO3H
I2142
KN-62, ≥95%, powder
C38H35N5O6S2
K1385
KN-93, ≥98% (HPLC)
C26H29ClN2O4S · H3PO4