EMAIL THIS PAGE TO A FRIEND
Beilstein journal of nanotechnology
Orthogonal chemical functionalization of patterned gold on silica surfaces.
PMID 26734519
Abstract
Single-step orthogonal chemical functionalization procedures have been developed with patterned gold on silica surfaces. Different combinations of a silane and a thiol were simultaneously deposited on a gold/silica heterogeneous substrate. The orthogonality of the functionalization (i.e., selective grafting of the thiol on the gold areas and the silane on the silica) was demonstrated by X-ray photoelectron spectroscopy (XPS) as well as time-of-flight secondary ion mass spectrometry (ToF-SIMS) mapping. The orthogonal functionalization was used to immobilize proteins onto gold nanostructures on a silica substrate, as demonstrated by atomic force microscopy (AFM). These results are especially promising in the development of future biosensors where the selective anchoring of target molecules onto nanostructured transducers (e.g., nanoplasmonic biosensors) is a major challenge.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
448931
Trichloro(1H,1H,2H,2H-perfluorooctyl)silane, 97%
C8H4Cl3F13Si