EMAIL THIS PAGE TO A FRIEND
Medicine
Short-term Follow-up US Leads to Higher False-positive Results Without Detection of Structural Recurrences in PTMC.
PMID 26735548
Abstract
To investigate the value of the annual follow-up neck ultrasonography (US) for postoperative surveillance in patients with papillary thyroid microcarcinoma (PTMC). This retrospective study has been approved by our institutional review board (IRB) with waiver for informed consent. A total of 375 patients diagnosed as PTMCs, who underwent total thyroidectomy with radioiodine remnant ablation were included, to identify the recurrence rate and the false-positive rate of annual ultrasound. The number, interval, and the results of follow-up US or fine needle aspiration were obtained from electronic medical records. Four (1.1%, 4/375) recurrences were found 3 years after the initial treatment, and only 1 patient (0.3%, 1/375) had a metastatic lymph node larger than 8 mm in the shortest diameter on US found 7.6 years after initial treatment with biochemical abnormalities. Cumulative risk of having at least 1 false-positive exam was 8.3% by the 8th US, and 8.1% by the 8-9 year follow-up. Cox multivariate regression showed shorter interval of follow-up US and presence of lymph node metastasis at initial surgery are independent predictors affecting the cumulative false-positive results (hazard ratio [HR], 0.60; 95% confidence interval [CI]: 0.49-0.73; P < 0.001 and HR, 2.19; 95% CI: 1.01-4.75; P = 0.048, respectively). Short-term follow-up US can result in higher cumulative false-positive results without detection of meaningful recurrences in patients with PTMCs who do not have biochemical abnormalities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76864
4-(4,5-Diphenyl-2-imidazolyl)phenol, suitable for chemiluminescence, ≥99.0% (HPLC)
C21H16N2O