EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
What are the differences between aerobic and anaerobic toxic effects of sulfonamides on Escherichia coli?
PMID 26748048
Abstract
Bacteria in the environment face the threat of antibiotics. However, most studies investigating the toxicity and toxicity mechanisms of antibiotics have been conducted on microorganisms in aerobic conditions, while studies examining the anaerobic toxicity and toxicity mechanisms of antibiotics are still limited. In this study, we determined the aerobic and anaerobic toxicities of sulfonamides (SAs) on Escherichia coli. Next, a comparison of the aerobic and anaerobic toxicities indicated that the SAs could be divided into three groups: Group I: log(1/EC50-anaerobic)>log(1/EC50-aerobic) (EC50-anaerobic/EC50-aerobic, the median effective concentration under anaerobic/aerobic conditions), Group II: log(1/EC50-anaerobic)≈log(1/EC50-aerobic), and Group III: log(1/EC50-anaerobic)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277053
(S)-(+)-2-(Methoxymethyl)pyrrolidine, 99%
C6H13NO