EMAIL THIS PAGE TO A FRIEND
Clinical nutrition (Edinburgh, Scotland)
Effect of parenteral infusion of fish oil-based lipid emulsion on systemic inflammatory cytokines and lung eicosanoid levels in experimental acute pancreatitis.
PMID 26758374
Abstract
Parenteral fish oil lipid emulsion (FOLE) might mitigate inflammation after injury. Acute pancreatitis (AP) can occur following major surgery and is characterized by tissue and systemic release of inflammatory mediators that contributes to the systemic inflammatory response syndrome and multiple organ failure. We evaluated the effect of short-term FOLE infusion before experimental induction of AP on systemic cytokine and lung eicosanoid profiles. Lewis rats (nxa0=xa072) received parenteral infusion of FOLE (FO group) or saline (SS group), or remained without parenteral infusion (CG group) for 48xa0h. Thereafter, AP was induced by retrograde injection of sodium taurocholate into the pancreatic duct. Animals were sacrificed after 2, 12 and 24xa0h. Blood and lung samples were collected to assess serum inflammatory cytokines (Luminex) and tissue eicosanoids (ELISA), respectively. Serum TNF-α increased over time and serum IL-10 decreased from 12 to 24xa0h in CG group. In SS group serum TNF-α increased from 12 to 24xa0h (pxa0=xa00.039) and serum IL-10 decreased over time. Both CG and SS groups exhibited increased IL-6/IL-10 ratio (pxa0=xa00.040). From 12 to 24xa0h animals from FO group showed decreased serum IL-1 (pxa0