EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
The TRAIL receptor agonist drozitumab targets basal B triple-negative breast cancer cells that express vimentin and Axl.
PMID 26759246
Abstract
Previously, we found that GST-tagged tumor necrosis factor-related apoptosis inducing ligand preferentially killed triple-negative breast cancer (TNBC) cells with a mesenchymal phenotype by activating death receptor 5 (DR5). The purpose of this study was to explore the sensitivity of breast cancer cell lines to drozitumab, a clinically tested DR5-specific agonist; identify potential biomarkers of drozitumab-sensitive breast cancer cells; and determine if those biomarkers were present in tumors from patients with TNBC. We evaluated viability, caspase activity, and sub-G1 DNA content in drozitumab-treated breast cancer cell lines and we characterized expression of potential biomarkers by immunoblot. Expression levels of vimentin and Axl were then explored in 177 TNBC samples from a publically available cDNA microarray dataset and by immunohistochemistry (IHC) in tumor tissue samples obtained from 53 African-American women with TNBC. Drozitumab-induced apoptosis in mesenchymal TNBC cell lines but not in cell lines from other breast cancer subtypes. The drozitumab-sensitive TNBC cell lines expressed the mesenchymal markers vimentin and Axl. Vimentin and Axl mRNA and protein were expressed in a subset of human TNBC tumors. By IHC, ~15 % of TNBC tumors had vimentin and Axl expression in the top quartile for both. These findings indicate that drozitumab-sensitive mesenchymal TNBC cells express vimentin and Axl, which can be identified in a subset of human TNBC tumors. Thus, vimentin and Axl may be useful to identify TNBC patients who would be most likely to benefit from a DR5 agonist.