EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Selective and Efficient Generation of ortho-Brominated para-Substituted Phenols in ACS-Grade Methanol.
PMID 26771597
Abstract
The mono ortho-bromination of phenolic building blocks by NBS has been achieved in short reaction times (15-20 min) using ACS-grade methanol as a solvent. The reactions can be conducted on phenol, naphthol and biphenol substrates, giving yields of >86% on gram scale. Excellent selectivity for the desired mono ortho-brominated products is achieved in the presence of 10 mol % para-TsOH, and the reaction is shown to be tolerant of a range of substituents, including CH3, F, and NHBoc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9143
Phorbol 12,13-diacetate, powder
C24H32O8