EMAIL THIS PAGE TO A FRIEND
Nature methods
An informatic framework for decoding protein complexes by top-down mass spectrometry.
PMID 26780093
Abstract
Efforts to map the human protein interactome have resulted in information about thousands of multi-protein assemblies housed in public repositories, but the molecular characterization and stoichiometry of their protein subunits remains largely unknown. Here, we report a computational search strategy that supports hierarchical top-down analysis for precise identification and scoring of multi-proteoform complexes by native mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3580
Toyocamycin, ≥98% (HPLC), from Streptomyces rimosus
C12H13N5O4