EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmaceutical sciences
Development and Validation of Stability-indicating High Performance Liquid Chromatographic Method for the Estimation of Everolimus in Tablets.
PMID 26798176
Abstract
The present study depicts the development of a validated reversed-phase high performance liquid chromatographic method for the determination of the everolimus in presence of degradation products or pharmaceutical excipients. Stress study was performed on everolimus and it was found that it degrade sufficiently in oxidizing and acidic conditions but less degradation was found in alkaline, neutral, thermal and photolytic conditions. The separation was carried out on Hypersil BDS C18 column (100×4.6 mm, 5 μ) column having particle size 5 μ using acetate buffer:acetonitrile (50:50 v/v) with pH 6.5 adjusted with orthophosphoric acid as mobile phase at flow rate of 1 ml/min. The wavelength of the detection was 280 nm. A retention time (Rt) nearly 3.110 min was observed. The calibration curve for everolimus was linear (r(2)=0.999) from range of 25-150 μg/ml with limit of detection and limit of quantification of 0.036 μg/ml and 0.109 μg/ml, respectively. Analytical validation parameters such as selectivity, specificity, linearity, accuracy and precision were evaluated and relative standard deviation value for all the key parameters were less than 2.0%. The recovery of the drug after standard addition was found to be 100.55%. Thus, the developed RP-HPLC method was found to be suitable for the determination of everolimus in tablets containing various excipients.