EMAIL THIS PAGE TO A FRIEND
Molecular cancer
miR-508-3p concordantly silences NFKB1 and RELA to inactivate canonical NF-κB signaling in gastric carcinogenesis.
PMID 26801246
Abstract
NF-κB signaling pathway plays an important role in gastric carcinogenesis. The basic expression and functional role of NFKB1 and RELA (components of canonical NF-κB pathway) in gastric cancer (GC) have not been well elucidated. In this study, the role of NFKB1 and RELA in gastric tumorigenesis will be investigated and their regulation by microRNAs (miRNAs) will be deeply explored. The mRNA and protein expression of NFKB1 and RELA were investigated by qRT-PCR and Western blot in GC cell lines and primary tumors. The functional roles of NFKB1 and RELA in GC were demonstrated by MTT proliferation assay, monolayer colony formation, cell invasion and migration, cell cycle analysis and in vivo study through siRNA mediated knockdown. Identification of NFKB1 as a direct target of tumor suppressor miRNA miR-508-3p was achieved by expression regulation assays together with dual luciferase activity experiments. NFKB1 and RELA were up-regulated in GC cell lines and primary tumors compared with normal gastric epithelium cells and their upregulation correlation with poor survival in GC. siRNA mediated knockdown of NFKB1 or RELA exhibited anti-oncogenic effect both in vitro and in vivo. NFKB1 was further revealed to be a direct target of miR-508-3p in gastric tumorigenesis and their expression showed negative correlation in primary GC samples. miR-508-3p was down-regulated in GC cells compared with normal gastric epithelium samples and its ectopic expression in GC cell lines also exerts tumor suppressor function. NFKB1 re-expression was found to partly abolish the tumor-suppressive effect of miR-508-3p in GC. All these findings supports that canonical NF-κB signaling pathway is activated in GC at least by the inactivation of miR-508-3p and this might have therapeutic potential in GC treatment.