EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Tongxinluo reduces brain edema and inhibits post-ischemic inflammation after middle cerebral artery occlusion in rats.
PMID 26805468
Abstract
Tongxinluo (TXL), a widely used traditional Chinese medicine, has been proved multiple therapeutic effects in cerebral ischemic infraction. The purpose of this study was to investigate the protective effects of TXL on the brain edema and post-ischemic inflammatory response. Middle cerebral artery occlusion in the rat was used as the ischemia model. Rats were treated with TXL. In the first stage, the best dosage was chosen based on functional assessment and infarct size. In the second stage, rats were randomly divided into 5 groups: sham control (sham), ischemia and reperfusion (IR) 24h, TXL24h, I/R72h, TXL72h. TXL(1.6g/kg/day) administration was pre-performed for 3 days in TXL groups, and was post-performed for 24h (TXL24h group) or 72h (TXL72h group). Brain edema was measured by water content, MRI and AQP4 expression. Iba1, HMGB1, TLR4, NF-κB expression were examined by immunofluorescence staining or Western blot. TNF-α was determined by enzyme-linked immunosorbent assay. High dose (1.6g/kg/day) of TXL remarkably reduced neurological deficit scores and cerebral infarct area. Compared with those results of I/R24h group, pre-post treatment with TXL for 3 days decreased brain water content, down-regulated AQP4 expression, lowered relative signal intensity of T2WI, reduced lesion volume ratio, and inhibited the activation of microglia, HMGB1, TLR4, NF-κB and TNF-α. These results indicated that the TXL pre-post treatment for 3 days could be an effective therapy for brain ischemia by inhibiting the development of brain edema and post-ischemic inflammation.