EMAIL THIS PAGE TO A FRIEND
Nature methods
Multi-target spectrally resolved fluorescence lifetime imaging microscopy.
PMID 26808668
Abstract
We introduce a pattern-matching technique for efficient identification of fluorophore ratios in complex multidimensional fluorescence signals using reference fluorescence decay and spectral signature patterns of individual fluorescent probes. Alternating pulsed laser excitation at three different wavelengths and time-resolved detection on 32 spectrally separated detection channels ensures efficient excitation of fluorophores and a maximum gain of fluorescence information. Using spectrally resolved fluorescence lifetime imaging microscopy (sFLIM), we were able to visualize up to nine different target molecules simultaneously in mouse C2C12 cells. By exploiting the sensitivity of fluorescence emission spectra and the lifetime of organic fluorophores on environmental factors, we carried out fluorescence imaging of three different target molecules in human U2OS cells with the same fluorophore. Our results demonstrate that sFLIM can be used for super-resolution multi-target imaging by stimulated emission depletion (STED).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T511285
5-Ethynyl-2′-deoxyuridine, (EdU), AldrichCPR
C11H12N2O5