EMAIL THIS PAGE TO A FRIEND
Mutation research
The genetic toxicology of benzoin and caprolactam.
PMID 2682234
Abstract
A summary is presented of the published literature on the genetic toxicology of the two rodent non-carcinogens benzoin and caprolactam.
Related Materials