EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Practical Ni-Catalyzed Aryl-Alkyl Cross-Coupling of Secondary Redox-Active Esters.
PMID 26835704
Abstract
A new transformation is presented that enables chemists to couple simple alkyl carboxylic acids with aryl zinc reagents under Ni-catalysis. The success of this reaction hinges on the unique use of redox-active esters that allow one to employ such derivatives as alkyl halides surrogates. The chemistry exhibits broad substrate scope and features a high degree of practicality. The simple procedure and extremely inexpensive nature of both the substrates and pre-catalyst (NiCl2·6H2O, ca. $9.5/mol) bode well for the immediate widespread adoption of this method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ALD00564
N-Hydroxytetrachlorophthalimide
C8HCl4NO3
Set your institution to read full text papers when available.