EMAIL THIS PAGE TO A FRIEND
Scientific reports
Metal ion transport quantified by ICP-MS in intact cells.
PMID 26838181
Abstract
The use of ICP-MS to measure metal ion content in biological tissues offers a highly sensitive means to study metal-dependent physiological processes. Here we describe the application of ICP-MS to measure membrane transport of Rb and K ions by the Na,K-ATPase in mouse skeletal muscles and human red blood cells. The ICP-MS method provides greater precision and statistical power than possible with conventional tracer flux methods. The method is widely applicable to studies of other metal ion transporters and metal-dependent processes in a range of cell types and conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L512087
PPT, AldrichCPR
C36H26O2P2S