EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of the Fatty Esters of Solketal and Glycerol-Formal: Biobased Specialty Chemicals.
PMID 26840287
Abstract
The caprylic, lauric, palmitic and stearic esters of solketal and glycerol formal were synthesized with high selectivity and in good yields by a solvent-free acid catalyzed procedure. No acetal hydrolysis was observed, notwithstanding the acidic reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122696
DL-1,2-Isopropylideneglycerol, ≥97.0%
C6H12O3