EMAIL THIS PAGE TO A FRIEND
Acta neurochirurgica
Restoration of ulnar nerve motor function by pronator quadratus motor branch: an anatomical study.
PMID 26860598
Abstract
The traditional surgical approach to repair of brachial plexus lesions involves use of whole segment ulnar nerve graft for contralateral seventh cervical (cC7) nerve root transfer, which sabotages the possibility of ulnar nerve recovery. We assessed the anatomical feasibility of a new approach that involves preservation of the motor branch of ulnar nerve (MBUN), for a later stage repair using the recovered pronator quadratus motor branch (PQMB), subsequent to the cC7 transfer procedure. Twenty-seven adult cadaver arms and one side of fresh adult cadaver were used in this study. The anterior interosseous nerve and its PQMB, as well as the motor and sensory branches of the ulnar nerve were dissected. The distances from the end of PQMB to the mid-point of a line joining the radial styloid and ulnar styloid, as well as to the point of divergence of the ulnar nerve, were measured. The MBUN was dissected from distal to proximal and the maximum length was measured. The diameter and number of axons of the nerve branches were also recorded. The distance from the end of the PQMB to the midpoint of the radial styloid and ulnar styloid was 6.04 ± 0.52xa0cm, and that to the point of divergence of the ulnar nerve was 8.02 ± 0.63xa0cm. The maximum length of the MBUN after its dissociation was 9.70 ± 1.38xa0cm. The mean diameters of axons of the MBUN and PQMB were 0.09 ± 0.02xa0cm and 0.05 ± 0.01xa0cm, respectively. The corresponding mean numbers of axons were 2913 ± 624 and 757 ± 183, respectively. The results indicate that the PQMB is suitable for transferring to the MBUN without nerve graft. This anatomical study paves the way for further testing of this new procedure after cC7 transfer in clinical settings.