EMAIL THIS PAGE TO A FRIEND
Medicine
Assessment of Arterial Stiffness, Volume, and Nutritional Status in Stable Renal Transplant Recipients.
PMID 26871855
Abstract
Reduction of cardiovascular death might have a significant effect on the long-term survival rates of renal transplant recipients (RTRs). The aim of the study was to assess the relation between arterial stiffness and graft function, adipose tissue content, and hydration status in patients after kidney transplantation (KTx).The study included 83 RTR patients (mean age: 55 ± 13 years) who had been admitted to a nephrology-transplantation outpatient clinic 0.5 to 24 years after KTx. Clinical and laboratory data were analyzed and eGFR was calculated with the CKD-EPI formula. Arterial stiffness was assessed in all RTRs with pulse wave propagation velocity (PWV) with the use of a complior device. In addition, fluid and nutritional status was assessed with a Tanita BC 418 body composition analyzer. The control group consisted of 31 hospital workers who received no medication and had no history of cardiovascular disease.Multivariable linear regression analysis, with PWV as a dependent variable, retained the following independent predictors in the final regression model: red blood cell distribution width (RDW) (B = 0.323; P = 0.004), age (B = 0.297; P = 0.005), tacrolimus therapy (B = -0.286; P = 0.004), and central DBP (B = 0.185; P = 0.041). Multivariable linear regression analysis with eGFR as a dependent variable retained the following independent predictors in the final regression model; creatinine concentration (B = -0.632; P = 0.000), hemoglobin (B = 0.280; P = 0.000), CRP (B = -0.172; P = 0.011), tacrolimus therapy (B = 0.142; P = 0.039), and triglycerides (B = -0.142; P = 0.035).Our data indicates that: kidney transplant recipients can present modifiable CVD risk factors linked to increased arterial stiffness, DBP, waist circumference, SCr, time on dialysis, CyA therapy, and visceral fat mass; RDW is a parameter associated with arterial stiffness; and parameters such as CyA therapy, time on dialysis, PWV, RDW, and triglycerides show negative associations with the allograft function assessed with eGFR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0531
BIO-Acetoxime, ≥98% (HPLC)
C18H12BrN3O3