EMAIL THIS PAGE TO A FRIEND
Animal reproduction science
Intracellular lipid dysregulation interferes with leukocyte function in the ovaries of meat-type hens under unrestricted feed intake.
PMID 26874430
Abstract
Meat-type Red-feather country hens fed ad libitum (AD-hens) exhibit obesity-associated morbidities and a number of ovarian irregularities. Leukocyte participations in ovarian activities are unstudied in AD-hens. In contrast to feed-restricted hens (R-hens), ovulatory process of the F1 follicle appeared delayed in AD-hens in association with reduced F1 follicle progesterone content, gelatinase A (MMP-2) and collagenase-3 (MMP-13) activities coincident with elevated IL-1β and no production (P<0.05), and increased leukocyte infiltration of inflamed necrotic follicle walls. Extracts of AD-hen F1 follicle walls induced greater leukocyte migration than extracts from F1 follicle wall extracts of R-hens (P<0.05). Co-cultures of granulosa cells with increasing numbers of leukocytes from either AD-hens or R-hens exhibited dose dependent reductions in progesterone production and increases in cell death. AD-hen leukocytes were less proapoptotic than their R counterparts (P<0.05). Granulosa MMP-13 and MMP-2 activities were also suppressed in the co-cultures with heterophils or monocytes in a dose-dependent manner (P<0.05). AD heterophils and R monocytes had a greater inhibitory effect on MMP activities in the co-cultures than their respective counterparts (P<0.05). Both basal and LPS-induced IL-1β secretion and MMP-22 or MMP-2 activities in freshly isolated AD-hen leukocytes were reduced (P<0.05). Exposure of AD or R leukocytes to 0.5mM palmitate impaired IL-1β secretion and MMP-22 or MMP-2 activity. Inhibition of ceramide synthesis with FB1 and ROS production with n-MPG scavenging rescued MMP activity and IL-1β production in palmitate treated heterophils, but exacerbated monocyte suppression. These latter findings suggest that intracellular lipid dysregulation in leukocytes contributes to ovarian dysfunction in AD-hens.
Related Materials