EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
CAV1 Prevents Gallbladder Cholesterol Crystallization by Regulating Biosynthesis and Transport of Bile Salts.
PMID 26875794
Abstract
Cholesterol gallstone disease (CGD) is a hepatobiliary disorder which results from a biochemical imbalance in the gallbladder bile. Here we show that loss of CAV1 sensitized mice to lithogenic diet-induced gallbladder cholesterol crystallization, which was associated with dysregulation of several hepatic transporters that efflux cholesterol, phospholipids, and bile salts. The combined effect of increased biliary cholesterol concentration and decreased biliary bile salt secretion in CAV1(-/-) mice led to an increased cholesterol saturation index and the formation of cholesterol crystals. At the signaling level, the ERK/AP-1 pathway seems to mediate the effects of CAV1 on biliary BA homeostasis and might be developed as a therapeutic target for CGD. We propose that CAV1 is an anti-lithogenic factor and that the CAV1(-/-) mice may offer a convenient CGD model to develop therapeutic interventions for this disease. J. Cell. Biochem. 117: 2118-2127, 2016. © 2016 Wiley Periodicals, Inc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5172
GW4064, ≥97% (HPLC)
C28H22Cl3NO4
723134
Tris(2-pyridylmethyl)amine, 98%
C18H18N4