EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[The unusual pain killing action of a widely used substance (gamma-butyrolactone)].
PMID 2687902
Abstract
An analgesic activity of gamma-butyrolactone was observed and described referring to a facilitated pain killing of certain diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90970
γ-Butyrolactone, analytical standard
C4H6O2