EMAIL THIS PAGE TO A FRIEND
Chemosphere
Adsorption of silver nanoparticles from aqueous solution on copper-based metal organic frameworks (HKUST-1).
PMID 26879292
Abstract
Silver nanoparticles (AgNP) are emerging pollutants. The use of novel materials such as Cu-(benzene 1,3,5-tricarboxylate, BTC) Metal-Organic Framework (MOFs), for AgNP adsorption and their removal from aqueous solutions has been studied. The effect of different parameters was followed and isotherm model was suggested. MOFs adsorbed fast and efficiently AgNP in the range C0 < 10 mg L(-1), being Freundlich isotherm (R = 0.993) these data fitted to. Among studied parameters a remarkable effect of chloride on sorption was found, thus their possible interactions were considered. The high adsorption efficiency of AgNP was achieved and it was found to be very fast. The feasibility of adsorption on Cu-(BTC) was proved in spiked waters. The results showed the potential interest of new material as adsorbent for removing AgNP from environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
688614
Basolite® C 300, produced by BASF
C18H6Cu3O12