EMAIL THIS PAGE TO A FRIEND
Scientific reports
Hyporesponsiveness to the anti-inflammatory action of interleukin-10 in type 2 diabetes.
PMID 26883847
Abstract
Chronic low-grade inflammation contributes to the pathology and complications of type 2 diabetes (T2D). Interleukin-10 (IL10), an anti-inflammatory cytokine, is suggested to play a protective role in T2D. However, the impact of T2D on IL10 function has not been previously assessed. We examined the ability of IL10 to inhibit inflammation in human T2D immune cells and explored underlying mechanisms using macrophage models. IL10 was less effective at inhibiting tumour necrosis factor (TNF)-α secretion in T2D whole blood cultures, which was not explained by altered IL10 receptor surface expression. These findings were observed in macrophages exposed to high glucose, which demonstrated similar IL10 resistance or hyporesponsiveness. These findings were also not explained by changes in IL10 receptor protein or other downstream signaling proteins. High glucose was also shown to impair the ability of IL10 to activate STAT3, a downstream signaling protein of IL10. Treatment with the SHIP1 agonist, AQX-MN100, reversed IL10 hyporesponsiveness in macrophages cultured in high glucose and showed equal effectiveness at different glucose conditions. This data supports the idea that IL10 hyporesponsiveness may contribute to chronic inflammation in T2D. These novel findings suggest that strategies aimed to overcome IL10 hyporesponsiveness may hold therapeutic potential for reducing inflammation in T2D.