EMAIL THIS PAGE TO A FRIEND
Scientific reports
PAX6 Isoforms, along with Reprogramming Factors, Differentially Regulate the Induction of Cornea-specific Genes.
PMID 26899008
Abstract
PAX6 is the key transcription factor involved in eye development in humans, but the differential functions of the two PAX6 isoforms, isoform-a and isoform-b, are largely unknown. To reveal their function in the corneal epithelium, PAX6 isoforms, along with reprogramming factors, were transduced into human non-ocular epithelial cells. Herein, we show that the two PAX6 isoforms differentially and cooperatively regulate the expression of genes specific to the structure and functions of the corneal epithelium, particularly keratin 3 (KRT3) and keratin 12 (KRT12). PAX6 isoform-a induced KRT3 expression by targeting its upstream region. KLF4 enhanced this induction. A combination of PAX6 isoform-b, KLF4, and OCT4 induced KRT12 expression. These new findings will contribute to furthering the understanding of the molecular basis of the corneal epithelium specific phenotype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1686
BIO, ≥98% (HPLC)
C16H10BrN3O2
SML1073
BIX, ≥98% (HPLC)
C9H7NO3S
R8279
RG108, ≥98% (HPLC), powder
C19H14N2O4