EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Beneficial Effects of Early Enteral Nutrition After Major Rectal Surgery: A Possible Role for Conditionally Essential Amino Acids? Results of a Randomized Clinical Trial.
PMID 26937858
Abstract
To investigate direct postoperative outcome and plasma amino acid concentrations in a study comparing early enteral nutrition versus early parenteral nutrition after major rectal surgery. Previously, it was shown that a low plasma glutamine concentration represents poor prognosis in ICU patients. A preplanned substudy of a previous prospective, randomized, open-label, single-centre study, comparing early enteral nutrition versus early parenteral nutrition in patients at high risk of postoperative ileus after surgery for locally advanced or locally recurrent rectal cancer. Early enteral nutrition reduced postoperative ileus, anastomotic leakage, and hospital stay. Tertiary referral centre for locally advanced and recurrent rectal cancer. A total of 123 patients with locally advanced or recurrent rectal carcinoma requiring major rectal surgery. Patients were randomized (ALEA web-based external randomization) preoperatively into two groups: early enteral nutrition (early enteral nutrition, intervention) by nasojejunal tube (n = 61) or early parenteral nutrition (early parenteral nutrition, control) by jugular vein catheter (n = 62). Eight hours after the surgical procedure artificial nutrition was started in hemodynamically stable patients, stimulating oral intake in both groups. Blood samples were collected to measure plasma glutamine, citrulline, and arginine concentrations using a validated ultra performance liquid chromatography-tandem mass spectrometric method. Baseline concentrations were comparable for both groups. Directly after rectal surgery, a decrease in plasma amino acids was observed. Plasma glutamine concentrations were higher in the parenteral group than in the enteral group on postoperative day 1 (p = 0.027) and day 5 (p = 0.008). Arginine concentrations were also significantly increased in the parenteral group at day 1 (p < 0.001) and day 5 (p = 0.001). Lower plasma glutamine and arginine concentrations were measured in the enteral group, whereas a better clinical outcome was observed. We conclude that plasma amino acids do not provide a causal explanation for the observed beneficial effects of early enteral feeding after major rectal surgery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36503
EPN, PESTANAL®, analytical standard
C14H14NO4PS