EMAIL THIS PAGE TO A FRIEND
Asian-Australasian journal of animal sciences
Effects of Direct-fed Microbial and Pine Cone Extract on Carcass Traits and Meat Quality of Hanwoo (Korean Native Cattle).
PMID 26954123
Abstract
The carcass traits and meat quality of Hanwoo (Korean native cattle) whose diets were supplemented with direct-fed microbial (DFM) and pine cone extract (PCE) were evaluated. Twenty head of Hanwoo steers were divided equally into four groups and for a period of 6 months were given different diets: One group was fed a basal diet as control (CON), the other three groups were fed a basal diet supplemented with DFM-1%, DFM+PCE-1% and DFM+PEC-3%, respectively. DFM+PCE3% diet resulted the lowest carcass quality grade. The loins of DFM-1% contained higher moisture and lower fat than did the loins from the CON group. The crude protein content of DFM+PCE-3% group was significantly higher than that of the other groups. The water holding capacity and Warner-Bratzler shear force of the DFM+PCE-1% and 3% groups were lower than those of the CON and DFM-1% groups. The DFM-1% and 3% groups contained lower saturated fatty acid, higher unsaturated fatty acid, mono-unsaturated fatty acid, and poly-unsaturated fatty acid than did CON and DFM+PCE 1% group. Moreover, the n6:n3 ratios of DFM-1% and DFM+PCE-1% and 3% groups were slightly lower than that of the CON group. Thus we concluded that DFM and PCE supplementation resulted healthier Hanwoo beef with lower fat content and n6:n3 ratio.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47015-U PUFA No.2 (Animal Source), analytical standard