EMAIL THIS PAGE TO A FRIEND
American journal of reproductive immunology (New York, N.Y. : 1989)
AGEs and HMGB1 Increase Inflammatory Cytokine Production from Human Placental Cells, Resulting in an Enhancement of Monocyte Migration.
PMID 26961863
Abstract
Advanced glycation end products (AGEs) and high-mobility group box-1 (HMGB1) are considered contributing to placental inflammation. We examined the effect of AGEs and HMGB1 on cytokines from Sw.71 human trophoblast cell lines and the interactions between Sw.71 cells and THP-1-monocytes. Sw.71 cells were cultured with/without AGEs or HMGB1. We examined the role of AGEs or HMGB1 on THP1 migration and effect of AGEs on IL-6 from Sw.71 cells using co-cultures or conditioned medium from THP-1 cells. AGEs and HMGB1 increased interleukin (IL)-6, IL-8, and chemokine C-C motif ligand 2 (CCL2) secretion from Sw.71 cells. The secretion of IL-6 was dependent on reactive oxygen species (ROS) and NF-κB. AGEs stimulated IL-6 secretion through receptor RAGE and TLR4, whereas HMGB1 stimulated it through TLR4. AGEs, but not HMGB1, increased monocyte migration via IL-8 and CCL2 from Sw.71 cells. THP-1 monocytes induced IL-6 secretion from Sw.71 cells, and AGEs further stimulated it. AGEs and HMGB1 may promote sterile placental inflammation cooperating with monocytes/macrophages.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2926
Diphenyleneiodonium chloride, ≥98%
C12H8ClI