EMAIL THIS PAGE TO A FRIEND
Oncotarget
Granulocyte-like myeloid derived suppressor cells (G-MDSC) are increased in multiple myeloma and are driven by dysfunctional mesenchymal stem cells (MSC).
PMID 26967390
Abstract
Granulocytic-Myeloid-derived suppressor cells (G-MDSC) are increased in Multiple Myeloma (MM) patients but the mechanisms of G-MDSC generation are still unknown. There are many evidences of the role of mesenchymal stem cells (MSC) in promoting MM cell growth, survival and drug-resistance. We here used a specific experimental model in vitro to evaluate the ability of MSC to induce G-MDSC. We found that although MSC derived from healthy donors (HD), MGUS and MM were able to generate the same amount of MDSC, only MM-MSC-educated G-MDSC exhibited suppressive ability. In addition, in comparison with MSC derived from HD, MM-MSC produce higher amount of immune-modulatory factors that could be involved in MDSC induction. Compared to G-MDSC obtained from co-culture models with MSC from healthy subjects, both MGUS and MM-MSC-educated G-MDSC showed increase of immune-modulatory factors. However, only MM-MSC educated G-MDSC 1) up-regulated immune-suppressive factors as ARG1 and TNFα, 2) expressed higher levels of PROK2, important in angiogenesis and inflammatory process, and 3) showed ability to digest bone matrix.Our data demonstrate that MM-MSC are functionally different from healthy subjects and MGUS-MSC, supporting an evolving concept regarding the contribution of MM-MSC to tumor development and progression.