EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
1,6-dinitropyrene.
PMID 2697765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9288
1,6-Dinitropyrene, ~60% (TLC)
C16H8N2O4
284327
1,6-Dinitropyrene, 98%
C16H8N2O4