PloS one
Set Configuration in Resistance Exercise: Muscle Fatigue and Cardiovascular Effects.
PMID 26982500
Abstract
Cardiovascular responses of traditional resistance (TS) training have been extensively explored. However, the fatigue mechanisms associated with an intra-set rest configuration (ISR) have not been investigated. This study compares two modalities of set configurations for resistance exercise that equates work to rest ratios and measures the central and peripheral fatigue in combination with cortical, hemodynamic and cardiovascular measures. 11 subjects performed two isometric knee extension training sessions using TS and ISR configurations. Voluntary activation (VA), single twitch amplitude, low frequency fatigue (LFF), Mwave, motor evoked potential (MEP), short intracortical inhibition (SICI), intracortical facilitation (ICF) and heart rate variability were evaluated before and after each training session. During each session beat to beat heart rate, blood pressure and rate pressure product (RPP) were also evaluated. After exercise VA decreased significantly for TS but not for ISR (P < 0.001), single twitch amplitude and LFF values were lower for TS than ISR (P < 0.004), and SICI was reduced only for the TS configuration (P = 0.049). During exercise RPP values were significantly higher for the TS than for ISR (P = 0.001). RPP correlated with VA for TS (r = -.85 P < 0.001) suggesting a relationship between central fatigue and cardiovascular stress. We conclude that ISR induced lower central and peripheral fatigue as well as lower cardiovascular stress in comparison with TS configuration. Our study suggests that set configuration is a key factor in the regulation of the neuromuscular and cardiovascular responses of resistance training.