EMAIL THIS PAGE TO A FRIEND
PeerJ
Palatability and pharmacokinetics of flunixin when administered to sheep through feed.
PMID 26989633
Abstract
Applying analgesics to feed is a potentially easy method of providing pain-relief to sheep and lambs that undergo painful husbandry procedures. To be effective, the medicated feed needs to be readily accepted by sheep and its consumption needs to result in therapeutic concentrations of the drug. In the present experiment, pelleted feed was supplemented with flunixin (4.0 mg/kg live weight) and offered to eight sheep. To test the palatability of flunixin, the individually penned sheep were offered normal feed and feed supplemented with flunixin in separate troughs for two consecutive days. A trend for a day by feed-type (control versus flunixin supplemented) interaction suggested that sheep may have had an initial mild aversion to pellets supplemented with flunixin on the first day of exposure, however, by on the second day there was no difference in consumption of normal feed and feed supplemented with flunixin. To test pharmacokinetics, sheep were offered 800 g of flunixin supplemented feed for a 12 h period. Blood samples were taken over 48 h and plasma drug concentrations were determined using ultra-high-pressure liquid chromatography, negative electrospray ionisation and tandem mass spectrometry. The mean ± S.D. time required to reach maximum concentration was 6.00 ± 4.14 h and ranged from 1 to 12 h. Average maximum plasma concentration was 1.78 ± 0.48 µg/mL and ranged from 1.61 to 2.80 µg/mL. The average half-life of flunixin was 7.95 ± 0.77 h and there was a mean residence time of 13.62 ± 1.17 h. Free access to flunixin supplemented feed enabled all sheep to obtain inferred therapeutic concentrations of flunixin in plasma within 6 h of starting to consume the feed. Provision of an analgesic in feed may be an alternative practical method for providing pain relief to sheep.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33586
Flunixin, VETRANAL, analytical standard
C14H11F3N2O2