EMAIL THIS PAGE TO A FRIEND
Head & neck
Prognostic value of FDG-PET volumetric parameters in patients with p16-positive oropharyngeal squamous cell carcinoma who received curative resection followed by postoperative radiotherapy or chemoradiotherapy.
PMID 27062085
Abstract
The purpose of this study was to determine whether pretreatment 18F-fluorodeoxyglucose-positron emission tomography (FDG-PET) volumetric parameters add more prognostic information to p16-positive oropharyngeal squamous cell carcinoma (SCC). We retrospectively analyzed 86 patients with p16-positive oropharyngeal SCC who underwent surgery and postoperative radiotherapy (RT) or chemoradiotherapy (CRT). Maximum standardized uptake value (SUVmax ), metabolic tumor volume (MTV), and total lesion glycolysis (TLG) were measured. Higher total MTV (tMTV), nodal MTV (nMTV), and tumor TLG (tTLG) were significantly associated with lower disease-free survival (DFS) (all p < .05) and higher distant metastasis rates (all p < .05). Multivariate analysis of DFS revealed that tMTV (p = .032), nMTV (p = .004), and tTLG (p = .018) were independently significant prognosticators. Patients with p16-positive disease with high metabolic tumor burden were associated with higher distant metastasis rates, translating into worse survival. These patients may not be optimal candidates for treatment deintensification. © 2016 Wiley Periodicals, Inc. Head Neck 38: First-1524, 2016.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F2756
Fluorescein di(β-D-galactopyranoside)
C32H32O15