EMAIL THIS PAGE TO A FRIEND
Contrast media & molecular imaging
Developing imidazoles as CEST MRI pH sensors.
PMID 27071959
Abstract
A series of intra-molecular hydrogen bonded imidazoles and related heterocyclic compounds were screened for their N-H chemical exchange saturation transfer (CEST) magnetic resonance imaging (MRI) contrast properties. Of the compounds, imidazole-4,5-dicarboxamides (I45DCs) were found to provide the strongest contrast, with the contrast produced at a large chemical shift from water (7.8 ppm) and strongly dependent on pH. We have tested several probes based on this scaffold, and demonstrated that these probes could be applied for in vivo detection of kidney pH after intravenous administration. Copyright © 2016 John Wiley & Sons, Ltd.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246115
4,5-Imidazoledicarboxylic acid, 97%
C5H4N2O4