EMAIL THIS PAGE TO A FRIEND
Food chemistry
Aliphatic glucosinolate synthesis and gene expression changes in gamma-irradiated cabbage.
PMID 27173540
Abstract
Glucosinolates, found principally in the plant order Brassicales, are modulated by different post-harvest processing operations. Among these, ionizing radiation, a non-thermal process, has gained considerable interest for ensuring food security and safety. In gamma-irradiated cabbage, enhanced sinigrin, a major glucosinolate, has been reported. However, the molecular basis of such a radiation induced effect is not known. Herein, the effect of radiation processing on the expression of glucosinolate biosynthetic genes was investigated. RT-PCR based expression analysis of seven glucosinolate biosynthetic pathway genes (MYB28, CYP79F1, CYP83A1, SUR1, UGT74B1, SOT18 and TGG1) showed that CYP83A1, MYB28, UGT74B1, CYP79F1 and SUR1 were up-regulated in irradiated cabbage. The content of jasmonates, signalling molecules involved in glucosinolate induction was, however, unaffected in irradiated cabbage suggesting their non-involvement in glucosinolate induction during radiation processing. This is the first report on the effect of gamma irradiation on the expression of glucosinolate biosynthetic genes in vegetables.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
333662
1H-1,2,3-Triazole, 97%
C2H3N3