EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Functional validation of tensin2 SH2-PTB domain by CRISPR/Cas9-mediated genome editing.
PMID 27246398
Abstract
Podocytes are terminally differentiated and highly specialized cells in the glomerulus, and they form a crucial component of the glomerular filtration barrier. The ICGN mouse is a model of glomerular dysfunction that shows gross morphological changes in the podocyte foot process, accompanied by proteinuria. Previously, we demonstrated that proteinuria in ICR-derived glomerulonephritis mouse ICGN mice might be caused by a deletion mutation in the tensin2 (Tns2) gene (designated Tns2(nph)). To test whether this mutation causes the mutant phenotype, we created knockout (KO) mice carrying a Tns2 protein deletion in the C-terminal Src homology and phosphotyrosine binding (SH2-PTB) domains (designated Tns2(ΔC)) via CRISPR/Cas9-mediated genome editing. Tns2(nph)/Tns2(ΔC) compound heterozygotes and Tns2(ΔC)/Tns2(ΔC) homozygous KO mice displayed podocyte abnormalities and massive proteinuria similar to ICGN mice, indicating that these two mutations are allelic. Further, this result suggests that the SH2-PTB domain of Tns2 is required for podocyte integrity. Tns2 knockdown in a mouse podocyte cell line significantly enhanced actin stress fiber formation and cell migration. Thus, this study provides evidence that alteration of actin remodeling resulting from Tns2 deficiency causes morphological changes in podocytes and subsequent proteinuria.
Related Materials