EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Monoterpene glycosides from Paeonia veitchii.
PMID 27268664
Abstract
The EtOH extract of the roots of Paeonia veitchii afforded two new monoterpene glycosides paeonidanin I (1) and paeonidanin J (2), and a new dimeric monoterpene glycoside paeonidanin K (3). Their structures were elucidated on the basis of spectroscopic means and hydrolysis products.