EMAIL THIS PAGE TO A FRIEND
Vascular
Midterm outcomes of catheter-directed interventions for the treatment of acute pulmonary embolism.
PMID 27271537
Abstract
Objective The hemodynamic benefits of catheter-directed thrombolysis for acute pulmonary embolism have not been clearly defined beyond the periprocedural period. The objective of this study is to report midterm outcomes of catheter-directed thrombolysis for treatment of acute pulmonary embolism. Methods Records of all patients undergoing catheter-directed thrombolysis for high- or intermediate-risk pulmonary embolism were retrospectively reviewed. Endpoints were clinical success, procedure-related complications, mortality, and longitudinal echocardiographic parameter improvement. Results A total of 69 patients underwent catheter-directed thrombolysis (mean age 59 ± 15 y, 56% male). Eleven had high-risk and 58 intermediate-risk pulmonary embolism. Baseline characteristics did not differ by pulmonary embolism subtype. Fifty-two percent of patients underwent ultrasound-assisted thrombolysis, 39% standard catheter-directed thrombolysis, and 9% other interventional therapy; 89.9% had bilateral treatment. Average treatment time was 17.7 ± 11.3 h with average t-Pa dose of 28.5 ± 19.6 mg. The rate of clinical success was 88%. There were two major (3%) and six minor (9%) periprocedural bleeding complications with no strokes. All echocardiographic parameters demonstrated significant improvement at one-year follow-up. Pulmonary embolism-related in-hospital mortality was 3.3%, and estimated survival was 81.2% at one year. Conclusions Catheter-directed thrombolysis is safe and effective for treatment of acute pulmonary embolism, with sustained hemodynamic improvement at one year. Further prospective large-scale studies are needed to determine comparative effectiveness of interventions for acute pulmonary embolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21861
1,1′-Carbonyl-di-(1,2,4-triazole), technical, ≥90% (T)
C5H4N6O